4-(3'-Fluorbenzyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8208-1G BB-8208-250M
COBBBB-8208-1GEA 341 EUR
COBBBB-8208-1G COBBBB-8208-250M
4-(3'-Fluorbenzyloxy)phenylboronsäure
4-(3'-Fluorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₂BFO₃
MDL: MFCD08276970
CAS-Nummer: 1072951-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR