2-Benzyloxy-4-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Benzyloxy)-4-fluorophenyl)boronic acid, 2-Benzyloxy-4-fluorobenzeneboronic acid

BB-2685-1G BB-2685-5G BB-2685-25G
COBBBB-2685-1GEA 49.7 EUR
COBBBB-2685-1G COBBBB-2685-5G COBBBB-2685-25G
2-Benzyloxy-4-fluorphenylboronsäure
2-Benzyloxy-4-fluorphenylboronsäure
Formel: C₁₃H₁₂BFO₃
Molecular Weight: 246.04 g/mol
MDL: MFCD03788402
CAS-Nummer: 848779-87-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR