3-Benzyloxy-4-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Benzyloxy)-4-fluorophenylboronic acid

BB-3963-5G BB-3963-1G
COBBBB-3963-5GEA 761 EUR
COBBBB-3963-5G COBBBB-3963-1G
3-Benzyloxy-4-fluorphenylboronsäure
3-Benzyloxy-4-fluorphenylboronsäure
Formel: C₁₃H₁₂BFO₃
Molecular Weight: 246.0400 g/mol
MDL: MFCD09475829
CAS-Nummer: 957034-74-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR