N-(4-Methoxybenzyl)-N-methylbenzolsulfonamid
Lieferant: COMBI-BLOCKS
OT-0512-5G
OT-0512-25G
OT-0512-1G
COBBOT-0512-5GEA
0
EUR
COBBOT-0512-5G
COBBOT-0512-25G
COBBOT-0512-1G
N-(4-Methoxybenzyl)-N-methylbenzolsulfonamid
N-(4-Methoxybenzyl)-N-methylbenzolsulfonamid
Formel:
C₁₅H₁₇NO₃S |
MDL:
MFCD08070819 CAS-Nummer: 915916-89-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...