(1S)-1,7,7-Trimethylnorborn-2-en-2-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: [(1S)-1,7,7-Trimethylbicyclo[2.2.1]hept-2-en-2-yl]boronic acid

BB-2567-1G BB-2567-250M
COBBBB-2567-1GEA 2180 EUR
COBBBB-2567-1G COBBBB-2567-250M
(1S)-1,7,7-Trimethylnorborn-2-en-2-boronsäure
(1S)-1,7,7-Trimethylnorborn-2-en-2-boronsäure
Formel: C₁₀H₁₇BO₂
Molecular Weight: 180,05 g/mol
MDL: MFCD08705231
CAS-Nummer: 871333-99-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR