3-Chinolinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Quinoline-3-boronic acid pinacolate, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline, Quinoline-3-boronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(3-quinolyl)-1,3,2-dioxaborolane

PN-2384-5G PN-2384-1G
COBBPN-2384-5GEA 520 EUR
COBBPN-2384-5G COBBPN-2384-1G
3-Chinolinboronsäurepinakolester
3-Chinolinboronsäurepinakolester
Formel: C₁₅H₁₈BNO₂
Molecular Weight: 255.1199 g/mol
MDL: MFCD05155218
CAS-Nummer: 171364-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR