6-Chinolinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Quinoline-6-boronic acid Pinacol ester, 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline, Quinoline-6-boronic acid, pinacol ester, 2-(Quinolin-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-5182-5G PN-5182-1G
COBBPN-5182-5GEA 409 EUR
COBBPN-5182-5G COBBPN-5182-1G
6-Chinolinboronsäurepinakolester
6-Chinolinboronsäurepinakolester
Formel: C₁₅H₁₈BNO₂
Molecular Weight: 255.1199 g/mol
MDL: MFCD06411327
CAS-Nummer: 406463-06-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR