4-Brom-6-chlorchinolin

Lieferant: COMBI-BLOCKS

HC-2216-1G HC-2216-250M
COBBHC-2216-1GEA 404 EUR
COBBHC-2216-1G COBBHC-2216-250M
4-Brom-6-chlorchinolin
4-Bromo-6-chloroquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242,5000 g/mol
MDL: MFCD11505112
CAS-Nummer: 1070879-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR