6-Brom-2-chlorchinolin

Lieferant: CHEMPUR

80-0009.500 80-0009.1
CHMP80-0009.500EA 0 EUR
CHMP80-0009.500 CHMP80-0009.1
6-Brom-2-chlorchinolin
6-Bromo-2-chloroquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242.50 g/mol
MDL: MFCD04115272
CAS-Nummer: 1810-71-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR