6-Brom-4-chlorchinolin

Lieferant: COMBI-BLOCKS

HC-2242-1G HC-2242-5G
COBBHC-2242-1GEA 25.8 EUR
COBBHC-2242-1G COBBHC-2242-5G
6-Brom-4-chlorchinolin
6-Bromo-4-chloroquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242,50 g/mol
MDL: MFCD00511001
CAS-Nummer: 65340-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR