3-Brom-2-chlor-6-nitrochinolin

Lieferant: CHEMPUR

80-0005.500 80-0005.1
CHMP80-0005.500EA 0 EUR
CHMP80-0005.500 CHMP80-0005.1
3-Brom-2-chlor-6-nitrochinolin
3-Bromo-2-chloro-6-nitroquinoline
Formel: C₉H₄BrClN₂O₂
MDL: MFCD04115268
CAS-Nummer: 296759-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR