Ethyl-8-brom-4-hydroxy-6-(trifluormethoxy)-3-chinolincarboxylat

Lieferant: COMBI-BLOCKS

HC-6938-1G HC-6938-250M
COBBHC-6938-1GEA 0 EUR
COBBHC-6938-1G COBBHC-6938-250M
Ethyl-8-brom-4-hydroxy-6-(trifluormethoxy)-3-chinolincarboxylat
Ethyl-8-brom-4-hydroxy-6-(trifluormethoxy)-3-chinolincarboxylat
Formel: C₁₃H₉BrF₃NO₄
MDL: MFCD11504930
CAS-Nummer: 1072944-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR