5-Brom-8-(trifluormethoxy)chinolin

Lieferant: COMBI-BLOCKS

HC-2032-5G HC-2032-1G
COBBHC-2032-5GEA 0 EUR
COBBHC-2032-5G COBBHC-2032-1G
5-Brom-8-(trifluormethoxy)chinolin
5-Bromo-8-(trifluoromethoxy)quinoline
Formel: C₁₀H₅BrF₃NO
MDL: MFCD11504881
CAS-Nummer: 1065074-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR