8-Brom-6-(trifluormethoxy)chinolin
Lieferant: COMBI-BLOCKS
PY-7354-1G
PY-7354-5G
COBBPY-7354-1GEA
404
EUR
COBBPY-7354-1G
COBBPY-7354-5G
8-Brom-6-(trifluormethoxy)chinolin
8-Bromo-6-(trifluoromethoxy)quinoline
Formel:
C₁₀H₅BrF₃NO Molecular Weight: 292,520 g/mol |
MDL:
MFCD09972204 CAS-Nummer: 1020253-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...