5-Amino-6-bromchinolin

Lieferant: COMBI-BLOCKS

Synonyme: 6-Bromoquinolin-5-amine

HC-2061-5G HC-2061-1G
COBBHC-2061-5GEA 1490 EUR
COBBHC-2061-5G COBBHC-2061-1G
5-Amino-6-bromchinolin
5-Amino-6-bromoquinoline
Formel: C₉H₇BrN₂
Molecular Weight: 223,0693 g/mol
MDL: MFCD11110424
CAS-Nummer: 50358-39-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR