5-Amino-8-bromchinolin
Lieferant: COMBI-BLOCKS
Synonyme:
8-Bromoquinolin-5-amine
HC-2064-5G
HC-2064-1G
COBBHC-2064-5GEA
671
EUR
COBBHC-2064-5G
COBBHC-2064-1G
5-Amino-8-bromchinolin
5-Amino-8-bromoquinoline
Formel:
C₉H₇BrN₂ Molecular Weight: 223,0693 g/mol |
MDL:
MFCD11110428 CAS-Nummer: 116632-58-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...