5,7-Dibrom-8-methoxychinolin

Lieferant: COMBI-BLOCKS

HC-6776-5G HC-6776-1G
COBBHC-6776-5GEA 1180 EUR
COBBHC-6776-5G COBBHC-6776-1G
5,7-Dibrom-8-methoxychinolin
5,7-Dibromo-8-methoxyquinoline
Formel: C₁₀H₈BrNO
MDL: MFCD11504924
CAS-Nummer: 17012-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR