5,8-Dibrom-6-methoxychinolin

Lieferant: COMBI-BLOCKS

PY-1135-5G PY-1135-1G
COBBPY-1135-5GEA 303 EUR
COBBPY-1135-5G COBBPY-1135-1G
5,8-Dibrom-6-methoxychinolin
5,8-Dibromo-6-methoxyquinoline
Formel: C₁₀H₈BrNO
MDL: MFCD12756472
CAS-Nummer: 1187386-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR