6-Brom-4-methoxychinolin

Lieferant: COMBI-BLOCKS

HC-2409-1G HC-2409-250M
COBBHC-2409-1GEA 345 EUR
COBBHC-2409-1G COBBHC-2409-250M
6-Brom-4-methoxychinolin
6-Bromo-4-methoxyquinoline
Formel: C₁₀H₈BrNO
Molecular Weight: 238,0806 g/mol
MDL: MFCD04036192
CAS-Nummer: 874792-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR