Methyl-4-chlor-7-methoxy-2-chinolincarboxylat

Lieferant: COMBI-BLOCKS

HC-2268-1G HC-2268-5G
COBBHC-2268-1GEA 303 EUR
COBBHC-2268-1G COBBHC-2268-5G
Methyl-4-chlor-7-methoxy-2-chinolincarboxylat
Methyl-4-chlor-7-methoxy-2-chinolincarboxylat
Formel: C₁₂H₁₀ClNO₃
MDL: MFCD11855871
CAS-Nummer: 1133115-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR