Methyl-6,8-dichlor-4-hydroxychinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-6953-1G HC-6953-5G
COBBHC-6953-1GEA 204 EUR
COBBHC-6953-1G COBBHC-6953-5G
Methyl-6,8-dichlor-4-hydroxychinolin-2-carboxylat
Methyl-6,8-dichlor-4-hydroxychinolin-2-carboxylat
Formel: C₁₁H₇Cl₂NO₃
MDL: MFCD11053815
CAS-Nummer: 1065074-55-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR