Ethyl-6,8-dichlor-4-hydroxychinolin-3-carboxylat

Lieferant: COMBI-BLOCKS

HC-2298-5G HC-2298-1G
COBBHC-2298-5GEA 612 EUR
COBBHC-2298-5G COBBHC-2298-1G
Ethyl-6,8-dichlor-4-hydroxychinolin-3-carboxylat
Ethyl-6,8-dichlor-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₉Cl₂NO₃
MDL: MFCD00173372
CAS-Nummer: 25771-89-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR