Methyl-4,7-dichlor-8-fluorchinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-2566-5G HC-2566-1G
COBBHC-2566-5GEA 451 EUR
COBBHC-2566-5G COBBHC-2566-1G
Methyl-4,7-dichlor-8-fluorchinolin-2-carboxylat
Methyl-4,7-dichlor-8-fluorchinolin-2-carboxylat
Formel: C₁₁H₆Cl₂FNO₂
Molecular Weight: 274.752 g/mol
MDL: MFCD12026029
CAS-Nummer: 1150164-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR