5-Amino-6-chlorchinolin

Lieferant: COMBI-BLOCKS

HC-2110-5G HC-2110-1G
COBBHC-2110-5GEA 692 EUR
COBBHC-2110-5G COBBHC-2110-1G
5-Amino-6-chlorchinolin
5-Amino-6-chloroquinoline
Formel: C₉H₇ClN₂
Molecular Weight: 178,6200 g/mol
MDL: MFCD02947117
CAS-Nummer: 341010-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR