5-Amino-8-chlorchinolin

Lieferant: COMBI-BLOCKS

HC-2108-5G HC-2108-1G
COBBHC-2108-5GEA 812 EUR
COBBHC-2108-5G COBBHC-2108-1G
5-Amino-8-chlorchinolin
5-Amino-8-chloroquinoline
Formel: C₉H₇ClN₂
Molecular Weight: 178,6 g/mol
MDL: MFCD09802113
CAS-Nummer: 75793-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR