8-Chlor-5-nitrochinolin

Lieferant: COMBI-BLOCKS

HC-2107-1G HC-2107-5G
COBBHC-2107-1GEA 303 EUR
COBBHC-2107-1G COBBHC-2107-5G
8-Chlor-5-nitrochinolin
8-Chloro-5-nitroquinoline
Formel: C₉H₅ClN₂O₂
Molecular Weight: 208,6000 g/mol
MDL: MFCD00466079
CAS-Nummer: 22539-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR