8-(Trifluormethoxy)chinolin-5-boronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(8-(Trifluoromethoxy)quinolin-5-yl)boronic acid
BB-5328-1G
BB-5328-250M
COBBBB-5328-1GEA
937
EUR
COBBBB-5328-1G
COBBBB-5328-250M
8-(Trifluormethoxy)chinolin-5-boronsäure
8-(Trifluormethoxy)chinolin-5-boronsäure
Formel:
C₁₀H₇BF₃NO₃ Molecular Weight: 256.97 g/mol |
MDL:
MFCD11504851 CAS-Nummer: 1072951-46-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...