8-Fluor-5-nitrochinolin

Lieferant: COMBI-BLOCKS

HC-2224-5G HC-2224-1G
COBBHC-2224-5GEA 1320 EUR
COBBHC-2224-5G COBBHC-2224-1G
8-Fluor-5-nitrochinolin
8-Fluoro-5-nitroquinoline
Formel: C₉H₅FN₂O₂
Molecular Weight: 192,1466 g/mol
MDL: MFCD09048716
CAS-Nummer: 94832-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR