6-Brom-4-chlor-2-methylchinolin

Lieferant: COMBI-BLOCKS

HC-2146-5G HC-2146-1G
COBBHC-2146-5GEA 1020 EUR
COBBHC-2146-5G COBBHC-2146-1G
6-Brom-4-chlor-2-methylchinolin
6-Bromo-4-chloro-2-methylquinoline
Formel: C₁₀H₇BrClN
Molecular Weight: 256,530 g/mol
MDL: MFCD00272373
CAS-Nummer: 53364-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR