3-Brom-4-hydroxy-2-(trifluormethyl)chinolin

Lieferant: COMBI-BLOCKS

HC-2166-1G HC-2166-5G
COBBHC-2166-1GEA 61.9 EUR
COBBHC-2166-1G COBBHC-2166-5G
3-Brom-4-hydroxy-2-(trifluormethyl)chinolin
3-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline
Formel: C₁₀H₅BrF₃NO
MDL: MFCD03407378
CAS-Nummer: 59108-47-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR