6-Brom-2-ethoxy-4-methylchinolin

Lieferant: COMBI-BLOCKS

HC-2575-1G HC-2575-5G
COBBHC-2575-1GEA 204 EUR
COBBHC-2575-1G COBBHC-2575-5G
6-Brom-2-ethoxy-4-methylchinolin
6-Bromo-2-ethoxy-4-methylquinoline
Formel: C₁₂H₁₂BrNO
MDL: MFCD12756431
CAS-Nummer: 1187385-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR