4-Brom-2,8-dimethylchinolin

Lieferant: COMBI-BLOCKS

HC-2867-5G HC-2867-1G
COBBHC-2867-5GEA 595 EUR
COBBHC-2867-5G COBBHC-2867-1G
4-Brom-2,8-dimethylchinolin
4-Bromo-2,8-dimethylquinoline
Formel: C₁₁H₁₀BrN
MDL: MFCD00272434
CAS-Nummer: 203506-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR