3-Brom-7-chlor-4-hydroxy-8-methyl-2-(trifluormethyl)chinolin

Lieferant: COMBI-BLOCKS

HC-2536-5G HC-2536-1G
COBBHC-2536-5GEA 451 EUR
COBBHC-2536-5G COBBHC-2536-1G
3-Brom-7-chlor-4-hydroxy-8-methyl-2-(trifluormethyl)chinolin
3-Bromo-7-chloro-4-hydroxy-8-methyl-2-(trifluoromethyl)quinoline
Formel: C₁₁H₆BrClF₃NO
MDL: MFCD11855925
CAS-Nummer: 59108-22-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR