2-Methoxy-6,8-dimethylchinolin

Lieferant: COMBI-BLOCKS

PY-1348-5G PY-1348-1G
COBBPY-1348-5GEA 0 EUR
COBBPY-1348-5G COBBPY-1348-1G
2-Methoxy-6,8-dimethylchinolin
2-Methoxy-6,8-dimethylchinolin
Formel: C₁₂H₁₃NO
MDL: MFCD15143628
CAS-Nummer: 861581-28-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR