2,4-Dichlor-3-phenylchinolin

Lieferant: CHEMPUR

BTB10856.250 BTB10856.1
CHMPBTB10856.250EA 0 EUR
CHMPBTB10856.250 MAYBBTB10856.1
2,4-Dichlor-3-phenylchinolin
2,4-Dichloro-3-phenylquinoline
Formel: C₁₅H₉Cl₂N
CAS-Nummer: 108832-15-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR