2,4-Dichlor-6-fluor-3-phenylchinolin

Lieferant: CHEMPUR

NRB02825.250 NRB02825.1
CHMPNRB02825.250EA 0 EUR
CHMPNRB02825.250 MAYBNRB02825.1
2,4-Dichlor-6-fluor-3-phenylchinolin
2,4-Dichloro-6-fluoro-3-phenylquinoline
Formel: C₁₅H₈Cl₂FN
MDL: MFCD00101151
CAS-Nummer: 274691-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR