1-Cyclohexenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1-Cyclohexeneboronic acid pinacol ester, 2-(Cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Cyclohexene-1-boronic acid pinacol ester

PN-2511-5G PN-2511-1G
COBBPN-2511-5GEA 324 EUR
COBBPN-2511-5G COBBPN-2511-1G
1-Cyclohexenylboronsäurepinakolester
1-Cyclohexenylboronsäurepinakolester
Formel: C₁₂H₂₁BO₂
Molecular Weight: 208.1049 g/mol
MDL: MFCD05663845
CAS-Nummer: 141091-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR