1-Cyclohexenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Cyclohexeneboronic acid pinacol ester, 2-(Cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Cyclohexene-1-boronic acid pinacol ester

M-Clarity™ Qualitätsstufe = MQ100

650277-1G 650277-500MG
SIAL650277-1GEA 226 EUR
SIAL650277-1G SIAL650277-500MG
1-Cyclohexenylboronsäurepinakolester, Sigma-Aldrich®
1-Cyclohexenylboronsäurepinakolester
Formel: C₁₂H₂₁BO₂
Molecular Weight: 208.1049 g/mol
MDL: MFCD05663845
CAS-Nummer: 141091-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR