1-Cyclopentenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Cyclopent-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopentene, 1-Cyclopenteneboronic acid Pinacol ester, 2-(1-Cyclopentenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-2510-5G PN-2510-1G
COBBPN-2510-5GEA 243 EUR
COBBPN-2510-5G COBBPN-2510-1G
1-Cyclopentenylboronsäurepinakolester
1-Cyclopentenylboronsäurepinakolester
Formel: C₁₁H₁₉BO₂
Molecular Weight: 194,0784 g/mol
MDL: MFCD08669391
CAS-Nummer: 287944-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR