4-Isochinolinboronsäureneopentylglycolester

Lieferant: COMBI-BLOCKS

Synonyme: Isoquinoline-4-boronic acid, 2,2-dimethylpropane-1,3-diol cyclic ester, 4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)isoquinoline

NE-2436-5G NE-2436-1G
COBBNE-2436-5GEA 1070 EUR
COBBNE-2436-5G COBBNE-2436-1G
4-Isochinolinboronsäureneopentylglycolester
4-Isochinolinboronsäureneopentylglycolester
Formel: C₁₄H₁₆BNO₂
Molecular Weight: 241.0933 g/mol
MDL: MFCD06659058
CAS-Nummer: 844891-01-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR