4-Isochinolinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinoline

PN-2436-1G PN-2436-5G
COBBPN-2436-1GEA 125 EUR
COBBPN-2436-1G COBBPN-2436-5G
4-Isochinolinboronsäurepinakolester
4-Isochinolinboronsäurepinakolester
Formel: C₁₅H₁₈BNO₂
Molecular Weight: 255.1199 g/mol
MDL: MFCD05663891
CAS-Nummer: 685103-98-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR