5-Brom-8-nitroisochinolin

Lieferant: COMBI-BLOCKS

HC-6910-5G HC-6910-1G HC-6910-25G
COBBHC-6910-5GEA 103 EUR
COBBHC-6910-5G COBBHC-6910-1G COBBHC-6910-25G
5-Brom-8-nitroisochinolin
5-Bromo-8-nitroisoquinoline
Formel: C₉H₅BrN₂O₂
Molecular Weight: 253.0500 g/mol
MDL: MFCD02091227
CAS-Nummer: 63927-23-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR