Isochinolin-5-carbaldehyd ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Isoquinoline-5-carboxaldehyde

L19636.MD L19636.03
L19636.MDEA 58.7 EUR
L19636.MD L19636.03
Isochinolin-5-carbaldehyd ≥97%
Isochinolin-5-carbaldehyd

Purity: 97% CAS No.: 80278-67-7 Molecular Formula: C10H7NO Formula Weight: 157.17 Melting Point: 113-116° Beilstein Reference: 112875

Formel: C₁₀H₇NO
Molecular Weight: 157.17 g/mol
Schmelzpunkt: 113…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03412483
CAS-Nummer: 80278-67-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR