Isochinolin-5-amin Hydrochlorid

Lieferant: COMBI-BLOCKS

PY-7289-1G PY-7289-5G
COBBPY-7289-1GEA 0 EUR
COBBPY-7289-1G COBBPY-7289-5G
Isochinolin-5-amin Hydrochlorid
Isochinolin-5-amin Hydrochlorid
Formel: C₉H₉ClN₂
Molecular Weight: 180.6342 g/mol
MDL: MFCD00035269
CAS-Nummer: 152814-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR