4-Pyridinboronsäure Hydrochlorid
Lieferant: COMBI-BLOCKS
Synonyme:
Pyridin-4-ylboronic acid hydrochloride
BB-3461-5G
BB-3461-25G
BB-3461-1G
COBBBB-3461-5GEA
0
EUR
COBBBB-3461-5G
COBBBB-3461-25G
COBBBB-3461-1G
4-Pyridinboronsäure Hydrochlorid
4-Pyridinboronsäure Hydrochlorid
Formel:
C₅H₇BClNO₂ |
MDL:
MFCD06201888 CAS-Nummer: 913835-65-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...