5-Brom-2-(dimethylamino)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-N,N-dimethylpyridin-2-amine

PY-1116-5G PY-1116-25G PY-1116-1G
COBBPY-1116-5GEA 114 EUR
COBBPY-1116-5G COBBPY-1116-25G COBBPY-1116-1G
5-Brom-2-(dimethylamino)pyridin
5-Brom-2-(dimethylamino)pyridin
Formel: C₇H₉BrN₂
Molecular Weight: 201,0600 g/mol
MDL: MFCD00099486
CAS-Nummer: 26163-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR