6-Brom-2-(isopropylamino)pyridin

Lieferant: COMBI-BLOCKS

HC-2589-1G HC-2589-5G
COBBHC-2589-1GEA 312 EUR
COBBHC-2589-1G COBBHC-2589-5G
6-Brom-2-(isopropylamino)pyridin
6-Brom-2-(isopropylamino)pyridin
Formel: C₈H₁₁BrN₂
MDL: MFCD12026036
CAS-Nummer: 89026-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR