6-Brom-2-(methylamino)pyridin

Lieferant: COMBI-BLOCKS

Synonyme: 6-Bromo-N-methyl-pyridin-2-amine, 6-Bromo-N-methylpyridin-2-amine

HC-6485-5G HC-6485-1G
COBBHC-6485-5GEA 613 EUR
COBBHC-6485-5G COBBHC-6485-1G
6-Brom-2-(methylamino)pyridin
6-Brom-2-(methylamino)pyridin
Formel: C₆H₇BrN₂
Molecular Weight: 187,0400 g/mol
MDL: MFCD09261221
CAS-Nummer: 89026-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR