5-Brom-2-(ethylamino)-3-nitropyridin

Lieferant: COMBI-BLOCKS

PY-7256-5G PY-7256-1G
COBBPY-7256-5GEA 375 EUR
COBBPY-7256-5G COBBPY-7256-1G
5-Brom-2-(ethylamino)-3-nitropyridin
5-Brom-2-(ethylamino)-3-nitropyridin
Formel: C₇H₈BrN₃O₂
MDL: MFCD08692126
CAS-Nummer: 26820-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR