5-Brom-2-(methylamino)-3-nitropyridin

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-N-methyl-3-nitropyridin-2-amine

PY-7257-5G PY-7257-1G
COBBPY-7257-5GEA 421 EUR
COBBPY-7257-5G COBBPY-7257-1G
5-Brom-2-(methylamino)-3-nitropyridin
5-Brom-2-(methylamino)-3-nitropyridin
Formel: C₆H₆BrN₃O₂
Molecular Weight: 232.03 g/mol
MDL: MFCD07375006
CAS-Nummer: 70232-59-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR